Anti-Heat Shock Protein 90, alpha Mouse Monoclonal Antibody [clone: 10F737]

Supplier: US Biological
H1834-55A3-50
USBIH1834-55A3-50EA 350 GBP
USBIH1834-55A3-50
Anti-Heat Shock Protein 90, alpha Mouse Monoclonal Antibody [clone: 10F737]
Antibodies
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: Heat Shock Protein 90, alpha
Clonality: monoclonal
Clone: 10F737
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR