Anti-Heat Shock Protein 70, P. falciparum Rabbit Polyclonal Antibody

Supplier: US Biological
H1830-95S-100
USBIH1830-95S-100EA 575 GBP
USBIH1830-95S-100
Anti-Heat Shock Protein 70, P. falciparum Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence:1:50; Western Blot:1:2000

Type: Primary
Antigen: Heat Shock Protein 70, P. falciparum
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Heat Shock Protein 70, P. falciparum
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Immunogen His-tagged and purified P. falciparum Hsp70 C-terminus.
Form liquid
Purification purified by Protein A chromatography
Concentration ~1 mg/ml
Cross adsorption No
Storage buffer PBS, pH 7,4, 0,09% sodium azide, 50% glycerol.
Storage temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR