Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 10C423]

Supplier: US Biological
G8900-01B
USBIG8900-01BEA 552 GBP
USBIG8900-01B
Anti-Gram Negative Endotoxin Mouse Monoclonal Antibody [clone: 10C423]
Antibodies
Type: Primary
Antigen: Gram Negative Endotoxin
Clonality: monoclonal
Clone: 10C423
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR