Anti-CD14 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 9L810]

Supplier: US Biological
C2265-55G
USBIC2265-55GEA 525 GBP
USBIC2265-55G
Anti-CD14 Mouse Monoclonal Antibody (Carboxyfluorescein) [clone: 9L810]
Antibodies
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: 9L810
Conjugation: Carboxyfluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR