Anti-Rabbit Serum Albumin Goat Polyclonal Antibody

Supplier: US Biological
A1327-70E
USBIA1327-70EEA 354 GBP
USBIA1327-70E
Anti-Rabbit Serum Albumin Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Rabbit Serum Albumin (Rabbit Serum Albumin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR