Anti-Adeno-Associated Virus 2, VP1, VP2, VP3 Rabbit Polyclonal Antibody

Supplier: US Biological
A0868-36N
USBIA0868-36NEA 1070 GBP
USBIA0868-36N
Anti-Adeno-Associated Virus 2, VP1, VP2, VP3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:200-1:500; Immunoprecipitation: 1:10; Immunofluorescence: 1:50-1:100

Type: Primary
Antigen: Adeno-Associated Virus 2, VP1, VP2, VP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Adeno-associated Virus 2, VP1, VP2, VP3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Format Serum
Form liquid
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR