Anti-TSC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
043343-APC
USBI043343-APCEA 828 GBP
USBI043343-APC
Anti-TSC2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TSC2
Antigen name tuberous sclerosis 2
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation APC (Allophycocyanin)
Reactivity Human
Host Rabbit
Isotype IgG
ELISA Yes
Modification phospho-Ser1096
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2, contains ~0,09% sodium azide
Storage temperature Cool
Shipping temperature Ice
Tested applications Dot Blot, Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR