Crystallising Dishes, with Spout
Supplier: TECHNICAL QUARTZ SOLUTIONS
75150 75151 75152 75153 75154 75155
TSLA75150EA 39.7 GBP
TSLA75150 TSLA75151 TSLA75152 TSLA75153 TSLA75154 TSLA75155
Crystallising Dishes, with Spout
Trays/Dishes/Pots Crystallising Dishes
Temperature-resistant crystallising dishes made from transparent quartz glass.
  • Excellent chemical resistance
  • High resistance against thermal shock
  • Made for tasks up to 1050 °C
  • High purity
The TQS quartz glass crystallising dishes are made for tasks up to 1050 °C (maximum limit 1280 °C). High chemical resistance and purity characterise these products. Quartz glass products have a long life for frequent use with temperatures up to 1050 °C, but the durability is dependant upon use and applications.
Order Now
SPECIFICATIONS
Environmentally Preferable