Combustion boats
Supplier: TECHNICAL QUARTZ SOLUTIONS
73375 73382 75140 75141 75142 75143 75144 75145 75146 75147
TSLA73375EA 87.7 GBP
TSLA73375 TSLA73382 TSLA75140 TSLA75141 TSLA75142 TSLA75143 TSLA75144 TSLA75145 TSLA75146 TSLA75147
Combustion boats
Boats Combustion Boats
Combustion boats, quartz, glazed, with or without handle.
  • Very good chemical resistance
  • Very good temperature resistance