Cleanroom vacuum cleaners, CR-1400WD and CR-1400D
Supplier: TIGER-VAC
113203B 113204B 113204C
141-1416EA 3120 GBP
141-1416 141-1417 TIGV113204C
Cleanroom vacuum cleaners, CR-1400WD and CR-1400D
Vacuum Cleaners
New generation cleanroom vacuum cleaners available in dry only or in wet and dry configurations. Designed specifically for the pharmaceutical, semiconductor and microelectronics industry.
  • Multiple stages filtration system; simple and reliable filter change
  • Two-stage by-pass motor provides powerful suction and quiet operation
  • The recovery tank is made of Type 316 stainless steel and is fully autoclavable (up to 121 °C) including the wheels
  • The stainless steel floater (liquid cut-off) is detachable for easy dismantling and maintenance; it is also
More Product Information