Industrial vacuum cleaner, TV-25L (DT) MFS HEPA WT
Supplier: TIGER-VAC
111447B2
TIGV111447B2EA 3520 GBP
TIGV111447B2
Industrial vacuum cleaner, TV-25L (DT) MFS HEPA WT
Vacuum Cleaners
Industrial vacuum cleaner for continuous duty. Designed specifically for the food and pharmaceutical industries. It is ideal for general cleaning and for process integration.
  • Multiple stages filtration system
  • Simple and reliable filter change
  • Regenerative blower with TEFC induction motor rated for continuous duty
  • Vertically mounted, carriage with suspended motor top (CSMT) for areas with space constraints
  • Equipped with a Manual Filter Shaker (MFS) System
  • Detachable recovery tank (DT) for easy disposal of recovered materials
  • Available polyliner and clip for
More Product Information