Cleanroom wipes, AlphaLite® II
Supplier: Texwipe
TX1091
TEXWTX1091EA 738 GBP
TEXWTX1091
Cleanroom wipes, AlphaLite® II
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipes are primarily intended for the usage as flat mop covers for cleaning and decontaminating floors. Made of non-woven polyester, they are especially suitable for cleaning and removing residues.
  • Knit polyester for applying disinfectants and biocides
  • High absorbency and excellent abrasion resistance
Suitable for use in ISO cleanroom class 3 - 8 environments.