Cleanroom wipes, AlphaLite®
Supplier: Texwipe
TX1008 TX1008B
115-0294EA 1160 GBP
115-0294 TEXWTX1008B
Cleanroom wipes, AlphaLite®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These lightweight, high quality wipes are constructed from 100% continuous-filament polyester.
  • Low particle generation and low extraction levels
  • Permits rigorous wiping with minimal release of fibers and particles
  • Available as stacked or bulk packaged
Ideal for spill control and general wiping in ISO Class 5 - 6 environments. For use in cleanroom environments that demand further reduction of particulate contamination. Also suitable for cleaning and burnishing magnetic media and other sensitive surfaces.
Colour: White
Packaging: Wipes areMore Product Information