Digital CO₂ measuring instrument with app connection, testo 535
Supplier: Testo
0563.0535
TEST0563.0535EA 453 GBP
TEST0563.0535
Digital CO₂ measuring instrument with app connection, testo 535
Data Loggers
The compact CO₂ measuring instrument testo 535 with practical App connection allows you to carry out this important measurement easily and efficiently and thus ensure comfort and a healthy indoor climate. Long-term stable infrared sensor technology, the measured values are reproducible and comparable even over longer periods of time. Ideal for recurring comparative measurements. Also supported by the time and point averaging directly in the measuring instrument. By the way, the testo Smart App displays the measured values as a table or graph, stores the results and documents them. This notMore Product Information