Infrared thermometer, testo 835-H1
Supplier: Testo
0560.8353
TEST0560.8353EA 351 GBP
TEST0560.8353
Infrared thermometer, testo 835-H1
Thermometers Digital Thermometers
This non contact thermometer measures surface temperature, moisture and dew point. It therefore helps to detect mold in buildings.
  • 4-point-laser shows the exact measuring zone, effectively preventing wrong measurements
  • 50:1 optics provide excellent surface temperature measurements even over greater distances
  • Easy-to-use: Menu guide with icons and joystick operation
  • Saves up to 200 measurements
  • Define individual alarm thresholds, acoustic and optical alarm
  • Min/max values
  • Backlit display
  • Can be connected to other temperature sensors (optionally available)
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable