Table/workbench, with optional add-on storage units or rear rail only
Supplier: TEKNOMEK
MTSSD1800X750S TSSD1200X600 TSSD1500X600 TLWB-RR15X75 TSSD1500X900 TSSD1800X600 TSSD1800X750 TSSD1800X900
TEKMMTSSD1800X750SEA 0 GBP
TEKMMTSSD1800X750S TEKMTSSD1200X600 TEKMTSSD1500X600 TEKMTLWB-RR15X75 TEKMTSSD1500X900 TEKMTSSD1800X600 TEKMTSSD1800X750 TEKMTSSD1800X900
Table/workbench, with optional add-on storage units or rear rail only
Furniture Tables and Workbenches
High quality, fully welded 304 grade stainless steel workbench with rear rail, sturdy, hygienic and built to last. Choice of mobile or static, with option to add-on storage units too.
  • Fully welded 304 grade stainless steel
  • Brushed dull polish finish
  • Fully welded, no construction or assembly required
  • Hidden extra framework to facilitate add-on units
  • Single rear rail for add-on unit compatibility
  • Space for extra storage under unit
304 grade stainless steel, Sturdy 40 mm box section frame, designed with hygiene in mind, Floor clearance to rear rail is 155More Product Information