Stainless Steel Writing Desk with Undershelf
Supplier: TEKNOMEK
TLWD-1S60X40
TEKMTLWD-1S60X40EA 0 GBP
TEKMTLWD-1S60X40
Stainless Steel Writing Desk with Undershelf
Furniture Tables and Workbenches
This stainless steel writing desks are the ideal workspace for any industry which count hygiene and longevity as priorities. They come with lockable hinged lids and a choice of stainless steel adjustable feet or stainless steel castors for the mobile version.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • Inclined top with lockable compartment
  • Available with stainless steel feet or stainless steel castors
This robust 304L-grade stainless steel writing desks come in both static and mobile options and with a brushed dull polish finish. The deskMore Product Information