Potassium iodate, Supelco®

Supplier: Merck
Warning

60386-50G-F
SUPL60386-50G-FEA 275 GBP
SUPL60386-50G-F
Potassium iodate, Supelco®
Potassium iodate
Formula: KlO₃
MW: 214 g/mol
Melting Pt: 560
Density: 3.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011406
CAS Number: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1 II
Merck Index: 13,07726

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR