Zinc chloride, granular USP, JP

Supplier: Spectrum Chemical
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

ZI105-12KG ZI105-2.5KG ZI105-50KGBL ZI105-100GM ZI105-500GM
SPCMZI105-12KGEA 2460 GBP
SPCMZI105-12KG SPCMZI105-2.5KG SPCMZI105-50KGBL SPCMZI105-100GM SPCMZI105-500GM
Zinc chloride, granular USP, JP
Zinc chloride

Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR