Sodium pyruvate 99, reagent

Supplier: Spectrum Chemical

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

SO193-500GM
SPCMSO193-500GMEA 987 GBP
SPCMSO193-500GM
Sodium pyruvate 99, reagent
Sodium pyruvate

Sodium Pyruvate, Reagent is most often added to the cell culture media as an extra source of energy. Sodium pyruvate also has a protective action against hydrogen peroxide. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Density: 1,718 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR