Tin(II) chloride dihydrate USP

Supplier: Spectrum Chemical
Danger

Synonyms: Stannous chloride dihydrate

S4004-200GM S4004-5KG
SPCMS4004-200GMEA 0 GBP
SPCMS4004-200GM SPCMS4004-5KG
Tin(II) chloride dihydrate USP
Tin (II) chloride dihydrate
Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8 III
Merck Index: 13,08861

Order Now

Specification Test Results

APPEARANCE
IDENTIFICATION (A) BLACKISH-GREY PRECIPITATE FORMS
IDENTIFICATION (B) YELLOWISH,FLOCCULENT PRECIPITATE IS FORMED
ANALYSIS 1: A YELLOWISH, FLOCCULENT PRICIPITATE FORMED
ANALYSIS 2: PRECIPITATE DISSOLVES, SOLUTION IS CLEAR AND COLORLESS
IDENTIFICATION (C) POSITIVE FOR CHLORIDE
ASSAY (SnCl2•2H2O) 98.0 - 102.0 %
SULFATE Max. 500 ppm
IRON Max. 100 ppm
LEAD (Pb) Max. 50 µg/g
APPEARANCE OF SOLUTION CLEAR AND COLORLESS
SUBSTANCES NOT PRECIPITATED BY THIOACETAMIDE Max. 0.2 %
ELEMENTAL IMPURITIES AS REPORTED
RESIDUAL SOLVENTS AS REPORTED


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR