D-(-)-N-Methylglucamine ≥99.0% USP

Supplier: Spectrum Chemical

Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol

M1122-25KGBL M1122-100GM M1122-2.5KG M1122-500GM
SPCMM1122-25KGBLEA 8350 GBP
SPCMM1122-25KGBL SPCMM1122-100GM SPCMM1122-2.5KG SPCMM1122-500GM
D-(-)-N-Methylglucamine ≥99.0% USP
D(-)-N-Methylglucamine

Meglumine, USP is used as a functional excipient where it acts as a counterion to enhance API stability and solubility.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
MW: 195.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004707
CAS Number: 6284-40-8
Merck Index: 12,06154

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Completeness and Color of Solution To Pass Test
Melting Range 128 - 132 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR