D-Glucono-1,5-lactone 99,0 USP

Supplier: Spectrum Chemical

Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone

G1165-1KG
SPCMG1165-1KGEA 322 GBP
SPCMG1165-1KG
D-Glucono-1,5-lactone 99,0 USP
D-Glucono-1,5-lactone

Gluconolactone, USP is used as a slow release acidifier excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 385
Melting Pt: 153
Storage Temperature: Ambient
MDL Number: MFCD00006647
CAS Number: 90-80-2
Merck Index: 13,04470

Order Now

Specification Test Results

Assay 99.0 - 101.0%
Lead (Pb) max. 0.001%
Heavy Metals max. 0.002%
Reducing Substances To Pass Test
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR