Dibutyl sebacate 98

Supplier: Spectrum Chemical
Warning

Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate

D2915-1LT
SPCMD2915-1LTEA 227 GBP
SPCMD2915-1LT
Dibutyl sebacate 98
Dibutyl sebacate

Dibutyl Sebacate, also known as DBS, is used as a plasticizer in production of plastics. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –11 °C
Density: 0,936
Storage Temperature: Ambient
MDL Number: MFCD00027218
CAS Number: 109-43-3
EINECS: 203-672-5

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR