Charcoal activated, powder USP

Supplier: Spectrum Chemical

Synonyms: Carbon activated, Activated carbon, Activated charcoal

CA191-12KG CA191-2.5KG CA191-45KGBL CA191-500GM
SPCMCA191-12KGEA 4390 GBP
SPCMCA191-12KG SPCMCA191-2.5KG SPCMCA191-45KGBL SPCMCA191-500GM
Charcoal activated, powder USP
Charcoal activated

Form: Powder

Formula: C
MW: 12.01 g/mol
Melting Pt: 3550
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2 III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR