Sorvall™ Legend™ Micro 17/21 Microcentrifuges, Ventilated/Refrigerated
Supplier: SORVALL
75002430 75002435 75002440
SORV75002430EA 0 GBP
SORV75002430 SORV75002435 SORV75002440
Sorvall™ Legend™ Micro 17/21 Microcentrifuges, Ventilated/Refrigerated
Centrifuges Microcentrifuges
These microcentrifuges support micro-volume protocols such as nucleic acid or protein lysate preparation and PCR reaction set-up - all in a remarkably small footprint.
  • Refrigerated versions offer cooling in just nine minutes
  • ClickSeal biocontainment rotor lid 'clicks' in place, ensuring securely contained samples
  • New PPSU lids have improved strength and chemical resistance
  • Intuitive controls and easy to read display simplifies operation
  • Sleek, space-saving footprint
  • Rugged design for years of dependable service
  • Quite performance provides safe, stress free
More Product Information