TriLink® CleanAmp® 7-Deaza-dGTP

Supplier: TriLink BioTechnologies

N-9515-10 N-9515-2
SOLUN-9515-10EA 566 GBP
SOLUN-9515-10 SOLUN-9515-2
TriLink® CleanAmp® 7-Deaza-dGTP
Nucleic Acid Reagents Nucleotides

CleanAmp® dNTPs are chemically-modified nucleotides designed to improve specificity, sensitivity, and robustness of your PCR.


  • Improves PCR specificity, sensitivity, and robustness
  • Provides an alternative approach to hot-start PCR without a need for a specially designed DNA polymerase
  • Grade: RUO


The CleanAmp® 7-deaza-dGTP: - Contains CleanAmp® 7-deaza deoxyguanosine 5’ triphosphate in a 2 µmol or 10 µmol format - Is ideal for GC-rich PCR (~60% GC) due to promoting the template’s strand separation by lowering its melting temperature - Enables hot-start PCR for specificity and sensitivity, with a thermolabile protecting group at the 3’ hydroxyl of the nucleotides (CleanAmp® dNTP benefit) - Has been QC analysed by AX-HPLC, RP-HPLC, 31P NMR, mass spec, and a standardised PCR assay


Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5b3da009117deecb3998d2dfe54bc3c4492bf31&vid=b5b3da009117deecb3998d2dfe54bc3c4492bf31&prid=web&clid=SAPDAM
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Application

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR