Detergents for Glassware Washers
Supplier: SMEG
DETERLQC2 ACIDGLP2 ACIDGLC2 DETERLQSP2 DETERLIQUIDI2 DETERLQI2
SMEGDETERLQC2EA 129 GBP
SMEGDETERLQC2 SMEGACIDGLP2 SMEGACIDGLC2 SMEGDETERLQSP2 SMEGDETERLIQUIDI2 SMEGDETERLQI2
Detergents for Glassware Washers
Cleaning Agents Detergents and Surface Cleansers
Detergents designed to optimise cleaning efficiency and the thermal disinfection phase for laboratory glassware cleaning.
  • For thorough instrument cleaning
  • Alkaline detergents for washing and neutralisers for the neutralisation stage
DETERGLASS
• Universal alkaline powder
• Detergent for automatic washing of laboratory glassware
• Contains sodium hydroxide, sodium metasilicate pentahydrate, sodium triphosphate pentabasic, triclosene sodium dihydrate
• 10 kg pack
DETERLIQUID C2
• Strong universal liquid alkaline detergent phosphate-free for washing steelMore Product Information