Multirack™ Jr.
Supplier: Simport Scientific
S610-13B S610-13G S610-13L S610-13O S610-13Y S610-16G S610-16L S610-16O S610-16Y S610-30B S610-30G S610-30L S610-30O S610-30Y
SIMPS610-13BEA 160 GBP
SIMPS610-13B SIMPS610-13G SIMPS610-13L SIMPS610-13O SIMPS610-13Y SIMPS610-16G SIMPS610-16L SIMPS610-16O SIMPS610-16Y SIMPS610-30B SIMPS610-30G SIMPS610-30L SIMPS610-30O SIMPS610-30Y
Multirack™ Jr.
Racks Tube Racks
These MultiRack™ Jr. are ideal for use in incubators, refrigerators, freezers, under lab hoods and om benchtops.
  • Made of acetal
  • Available in three models for 13 mm, 16 mm and 25 to 30 mm tubes
  • Compact, lightweight, stackable and submersible
  • 3-tier facilitates stability of tubes
  • Base has rounded wells with drain holes
A newly designed tube support that can be used all around the lab. It is available in three models to accommodate a full range of laboratory test tubes and centrifuge tubes up to 30 mm in diameter. Although it is one of the mostMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable