Microarray molds kits, T-Sue™
Supplier: Simport Scientific
M473-15 M473-P30P M473-150 M473-36 M473-60 M473-72 M473-P20R M473-P10MA M473-150PC M473-15PC M473-170 M473-170PC M473-24 M473-24PC M473-36PC M473-60PC M473-72PC M473-P15Y M473-P40B
SIMPM473-15EA 1080 GBP
SIMPM473-15 SIMPM473-P30P SIMPM473-150 SIMPM473-36 SIMPM473-60 SIMPM473-72 SIMPM473-P20R SIMPM473-P10MA SIMPM473-150PC SIMPM473-15PC SIMPM473-170 SIMPM473-170PC SIMPM473-24 SIMPM473-24PC SIMPM473-36PC SIMPM473-60PC SIMPM473-72PC SIMPM473-P15Y SIMPM473-P40B
Microarray molds kits, T-Sue™
Microarray Instruments
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
  • Can be reused hundreds of times without losing its flexibility
  • Withstands temperatures between –100 to +250 °C
  • View many different samples on the same slide
  • Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-madeMore Product Information