Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Supplier: Industrial Physics
LD3059
SIMELD3059EA 468 GBP
SIMELD3059
Reference guide and photographs for steel surfaces, SSPC-VIS 5/NACE VIS 9
Books
This pictorial standard shows two uncoated, rusted steel surfaces (Conditions C and D) before and after wet abrasive blast cleaning. Photographs illustrate two degrees of cleaning (WAB 6 and WAB 10) for each initial condition with additional photos that depict the appearance of light, moderate, and heavy flash rust after cleaning. Also contains a written guide to the use of reference photographs and additional explanatory notes. Cleanliness requirements for conditions depicted are defined by the joint SSPC/NACE surface preparation specifications for commercial (SP 6/NACE 3) and near-whiteMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable