BalanCD™ CHO Feed 3, Powder
Supplier: Shenandoah Biotechnology
94118-10L
SHBT94118-10LEA 2370 GBP
SHBT94118-10L
BalanCD™ CHO Feed 3, Powder
Cell Culture Media
BalanCD™ CHO Feed 3 is a chemically-defined, animal component-free feed medium designed to increase process yields of antibodies and recombinant proteins in Chinese Hamster Ovary (CHO) cells in fed-batch mode.
This formulation was developed using FUJIFILM Irvine Scientific’s Rational Culture Media Design strategy to achieve enhanced performance of growth and production in fed-batch culture. BalanCD™ CHO Feed 3 was designed to be used with a wide range of growth media for optimal performance in fed-batch cultures.