HyClone™ Fetal Bovine Serum, Characterized (New Zealand), HyClone Products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30406.02 SH30406.02HI SH30406.02IR30 SH30406.02IH25 SH30406.02IR25 SH30406.02-45 SH30406.02IR35
HYCLSH30406.02EA 1430 GBP
HYCLSH30406.02 HYCLSH30406.02HI SH30406.02IR30 SH30406.02IH25 SH30406.02IR25 SH30406.02-45 SH30406.02IR35
HyClone™ Fetal Bovine Serum, Characterized (New Zealand), HyClone Products (Cytiva)
Sera (Serum)
HyClone™ Characterised FBS (New Zealand origin) is widely used by users requiring an extensive biochemical profile and full traceability.
  • Sourced in New Zealand with complete traceability back to the original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0,1 µm) pore size-rated filters
  • Virus panel testing according to 9 CFR 113,53
HyClone Characterised Fetal Bovine Serum (FBS), New Zealand Origin is carefullyMore Product Information