Calf serum, iron supplemented, US origin, HyClone™
Supplier: HyClone products (Cytiva)
SH30072.03 SH30072.02 SH30072.04 SH30072.02HI SH30072.03HI SH30072.04HI SH30072.03IR25 SH30072.04IH25 SH30072.04IR25 SH30072.04IR30
HYCLSH30072.03EA 94.1 GBP
HYCLSH30072.03 HYCLSH30072.02 HYCLSH30072.04 HYCLSH30072.02HI HYCLSH30072.03HI HYCLSH30072.04HI SH30072.03IR25 SH30072.04IH25 SH30072.04IR25 SH30072.04IR30
Calf serum, iron supplemented, US origin, HyClone™
Sera (Serum)
HyClone™ iron-supplemented bovine calf serum contains additional iron to boost serum transferrin levels to physiological levels.
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16 to 24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Supplemented with iron and growth-promoting factors
HyClone Iron-Supplemented Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves,More Product Information