Powder funnels, PP
Supplier: SEMADENI
0211 0213
SEMA0211EA 12 GBP
SEMA0211 SEMA0213
Powder funnels, PP
Funnels Powder Funnels
Funnels, made of PP, translucent. Used for transferring powders. Also suitable for liquids.
  • Parallel stem minimises powder bridging
  • Very good chemical and temperature resistance
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable