Universal waste hubs 'Jan'
Supplier: S.C.A.T.
320045 320050 320051 320053 320055 320060 320070 320063 320065 320083 320090 320095 420045 420060 420160 320160
SCTV320045EA 475 GBP
SCTV320045 SCTV320050 SCTV320051 SCTV320053 SCTV320055 SCTV320060 SCTV320070 SCTV320063 SCTV320065 SCTV320083 SCTV320090 SCTV320095 SCTV420045 SCTV420060 SCTV420160 SCTV320160
Universal waste hubs 'Jan'
Funnels Safety Funnels
Collect your laboratory waste with our 'Jan' Universal Waste Hub, the smart all-in-one solution for liquid waste. It is a clever combination of lid funnel with tubing and HPLC connections plus an integrated exhaust air filter. Many other innovative details guarantee maximum safety compared to conventional solutions.
  • High quality PE-HD and PTFE materials for maximum durability
  • Stable funnel lid with integrated sealing lip against evaporation
  • Removable dirt sieve for easy cleaning
  • Funnel basin (140 mm) with high flow rate for larger disposal volumes
  • Includes
More Product Information