Screw caps, YOUTILITY™ GL 45, DURAN®
Supplier: DWK Life Sciences
292292827
SCOT292292827EA 43.9 GBP
SCOT292292827
Screw caps, YOUTILITY™ GL 45, DURAN®
Closures Bottle Closures
The ergonomic-shaped YOUTILITY™ GL 45 screw cap features a liner less design compatible with both DURAN® original GL 45 bottles and YOUTILITY™ bottles. The cap is moulded from biocomptatible materials making it suitable for use in pharmaceutical manufacturing.
  • Certificate of Conformance (CoC)
  • Ergonomic shape with a comfortable grip
  • Autoclavable at 121 °C (20 minutes) or 134 °C (three minutes)
  • Max. usage temperature 140 °C
  • Inverted cone shape for a safer grip
The white YOUTILITY™ caps are retrace coded for traceability. The retrace code can be used toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable