Adapters, receiver, vacuum, bent, DURAN®
Supplier: DWK Life Sciences
241302105 241303401 241304603
SCOT241302105EA 70.7 GBP
SCOT241302105 SCOT241303401 SCOT241304603
Adapters, receiver, vacuum, bent, DURAN®
Distillation Components Distillation Adapters
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with two standard ground joints and screw-on plastic hose connection.
Order Now
SPECIFICATIONS
Environmentally Preferable