TitriSoft 3.5 - PC titration software for TITRONIC® 300, TITRONIC® 500, TitroLine® 7000 – 7800 and sample changer
Supplier: SI Analytics
285217950 285217960 285217970 285217980 285217990
SCOI285217950EA 3970 GBP
SCOI285217950 SCOI285217960 SCOI285217970 SCOI285217980 SCOI285217990
TitriSoft 3.5 - PC titration software for TITRONIC® 300, TITRONIC® 500, TitroLine® 7000 – 7800 and sample changer
Software Laboratory Software
The TitriSoft 3.3 titration software is the optimum solution for your titration tasks. The software can be used with Windows 7, 8.1 and 10 and supports your daily work procedures during sample preparation, titration and evaluation of the results. The software has been developed to be clear, logical and user-friendly.
In combination with the TitriSoft 3.5 / 3.5 P, a so-called 'parallel' titration can be carried out with a TitroLine® 7800 (also 7000 and 7750) and a TITRONIC®300 / 500 piston burette.
This means you only need one titrator and one piston burette to carry out two titrationsMore Product Information