Precision thermometers, for block thermostats, stem type
Supplier: Ludwig Schneider GmbH & CO. KG
64304 64303
SCNL64304EA 51 GBP
SCNL64304 SCNL64303
Precision thermometers, for block thermostats, stem type
Thermometers Analogue Thermometers
These thermometers are used for block thermostats, supplied in a plastic reaction tube, filled with 1 ml mineral oil.
  • Round, white back capillary form
  • Permanent graduations and lettering
  • Serial number and certificate (conformance and accuracy)
Blue filling, stem type.
Packaging: Single packed in a cardboard box.