Syringe filters, Minisart® Acticosart
Supplier: Sartorius
17840-Q
SART17840-QEA 2510 GBP
SART17840-Q
Syringe filters, Minisart® Acticosart
Filters Syringe Filters
The Minisart® Acticosart syringe filter with chemically inert hydrophobic PTFE is suitable for venting as well as filtration of gases with a pH of 3 to 12.
  • MBS housing suitable for glueing
  • PVC-free
  • 100% optical integrity test
  • Colour coded MBS housing for easy pore identification
  • Superior flow rate
Due to its hydrophobic nature the PTFE membrane functions as a water barrier and does not become wetted by aerosols. This keeps the hydrophobic venting filter permeable for gas or air. The active carbon filling adsorbs molecules. The housing of the filter isMore Product Information