Sartobind® Filtration Capsules
Supplier: Sartorius
96HICP42EUC11A 92IEXQ42D4-OOA 92STPA42DD-11D 92IEXQ42D4-SSA 92IEXS42D4-OOA 92IEXS42D4-SSA 92IEXQ42DD-11D 92HICP42DD-11D
SARP96HICP42EUC11AEA 689 GBP
SARP96HICP42EUC11A SARP92IEXQ42D4-OOA SARP92STPA42DD-11D SARP92IEXQ42D4-SSA SARP92IEXS42D4-OOA SARP92IEXS42D4-SSA SARP92IEXQ42DD-11D SARP92HICP42DD-11D
Sartobind® Filtration Capsules
Filters Membrane Filter Capsules
Ready to use disposable ion exchange capsules for capturing and polishing in biomanufacturing. Ideal for applications such as the removal of DNA, viruses, endotoxins and protein contaminants.
  • Simple handling
  • Fast operation
  • Less buffer consumption
Stabilised reinforced cellulose.
Order Now
SPECIFICATIONS
Type Sartobind® Phenyl Nano Sartobind® Q SingleSep Mini Sartobind STIC® PA Pico Sartobind® Q SingleSep Mini Sartobind® S SingleSep Mini Sartobind® S SingleSep Mini Sartobind® Q Pico Sartobind® Phenyl Pico
Environmentally Preferable
Water flow rate 15 ml/min 140 ml/min 0,8 - 2,4 ml/min 140 ml/min 140 ml/min 140 ml/min 0,8 - 2,4 ml/min 0,8 - 2,4 ml/min
Filtration area 110
Inlet/outlet Luer female connectors Hose barb connectors Luer female connectors Sanitary 1/2" - 3/4" Hose barb connectors Sanitary 1/2" - 3/4" Luer female connectors Luer female connectors
Ø×L 33×37 mm 45×103 mm 11×31 mm 45×87 mm 45×103 mm 45×87 mm 11×31 mm 11×31 mm
Max. pressure 4 bar 4 bar 6 bar 4 bar 4 bar 4 bar 6 bar 6 bar