Filtration capsules, Sartopore® 2 XLM
Supplier: Sartorius
5448307H3G-SS 5445358M8-OO-A
SARP5448307H3G-SSEA 1830 GBP
SARP5448307H3G-SS SARP5445358M8-OO-A
Filtration capsules, Sartopore® 2 XLM
Filters Membrane Filter Capsules
Sartopore® 2 XLM is designed with finer filter membranes (0,1 µm) for the successful retention of Mycoplasma. The build in 0,2 µm prefilter layer is specifically designed to protect the final 0,1 μm membrane layer in cell culture media filtration applications.
  • Very high effective filtration area per filter element allows for >50% more performance
  • Available in a broad range of sizes and formats to provide linear scale-up from R and D to process scale
PES membrane; support fleece: polyester/PP; core and end caps: PP; O-rings: silicone; housing: PP;More Product Information