ProfiLine OEM Weigh Cell
Supplier: Sartorius Balances
WZA224-N
SARBWZA224-NEA 9920 GBP
SARBWZA224-N
ProfiLine OEM Weigh Cell
Balances and Scales
The WZA-N series is the successor of the well established WZA-line. This series focus on weighing resolutions as low as 0,1 mg and capacities of up to 8200 g.
  • A monolithic weighing system in stainless steel IP 44 housing
  • Aluminium electronic box, all connection via the front
  • Force compensation technology weighing system with a resolution of 0,1 mg
  • Excellent weighing technology bundled with easy adaptation
  • Adjustable load receptor with overload protection, optional lift-off protection
  • Possibility for under floor weighing
  • LED's to control power and data
More Product Information