Cubis II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Supplier: Sartorius Balances
Cubis II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.