Sodium dihydrogen phosphate monohydrate ≥98% ACS, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

S9638-1KG S9638-6X1KG
S9638-1KGEA 96.1 GBP
S9638-1KG S9638-6X1KG
Sodium dihydrogen phosphate monohydrate ≥98% ACS, Fluka™
Sodium dihydrogen phosphate monohydrate
Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 650 °C (anhydr.)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
ADR: 3
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
pH (5 %, 25 °C) 4.1 - 4.5
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 50 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 100 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR