Anti-IgG (H+L) Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-9102
ROCK612-9102EA 248 GBP
ROCK612-9102
Anti-IgG (H+L) Chicken Polyclonal Antibody
Antibodies
Secondary Chicken Anti-IgG (H&L) Reacts with Rat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgG
Reactivity: Rat
Order Now

SPECIFICATIONS

Antigen symbol IgG (H+L)
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Rat
Host Chicken
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Immunogen Rat IgG whole molecule
Format Antibody fraction
Form Liquid (sterile filtered)
Purification purified by immunoaffinity chromatography
Concentration 2,0 mg/ml by UV absorbance at 280 nm
Cross adsorption none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR