Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Rockland Immunochemicals
609-1506
ROCK609-1506EA 360 GBP
ROCK609-1506
Anti-IgA Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
This product has been assayed against 1.0 µg of Human IgA in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgA α
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgA
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IgA
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation AP (Alkaline Phosphatase)
Reactivity Human
Host Goat
Isotype
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Gel shift Yes
Flow cytometry Yes
ChIP Yes
Neutralisation Yes
Immunogen Human IgA alpha heavy chain
Format Antibody fraction
Form Liquid (sterile filtered)
Purification purified by immunoaffinity chromatography
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Cross adsorption none
Storage buffer 0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR