Anti-COL1A1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
600-406-103
ROCK600-406-103EA 801 GBP
ROCK600-406-103
Anti-COL1A1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Anti-COLLAGEN Type I Antibody Biotin Conjugated is suitable for western blot, immunoprecipitatioin, Flow Cytometry, and immunochemistry. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:3000 - 1:6000; Western Blot: 1:3000 - 1:6000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Rat, Mouse, Mammalian
Order Now

SPECIFICATIONS

Antigen symbol COL1A1
Environmentally Preferable
Antigen name collagen, type I, alpha 1
Antigen synonyms EDSC|OI4|OI2|OI3|OI1
Antibody type Primary
Clonality Polyclonal
Conjugation Biotin
Reactivity Mammals,Human,Bovine,Rat,Mouse
Host Rabbit
Gene ID 1277
Isotype
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Immunogen Collagen Type I from human and bovine placenta.
Format Antibody fraction
Form Lyophilized
Purification purified by immunoaffinity chromatography
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Cross adsorption none
Storage buffer 0,125 M Sodium Borate, 0,075 M Sodium Chloride, 0,005 M EDTA, pH 8.0, 0,01% (w/v) Sodium Azide, Immunoglobulin and Protease free
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR