Anti-IRS1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-445
ROCK600-401-445EA 768 GBP
ROCK600-401-445
Anti-IRS1 Rabbit Polyclonal Antibody
Antibodies
This affinity purified antibody has been tested for use in ELISA and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 130 kDa in size corresponding to phosphorylated IRS1 Protein by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:250 - 1:1500;

Type: Primary
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IRS1
Antigen name insulin receptor substrate 1
Environmentally Preferable
Antigen synonyms HIRS-1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 3667
Isotype
Western blot Yes
ELISA Yes
ImmunoPrecipitation Yes
Modification phospho-Ser307
Epitope internal
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 298-316 of human IRS1 Protein.
Amino acid number 298 to 316
Format Antibody fraction
Form Liquid (sterile filtered)
Purification affinity-purified
Concentration 1,0 mg/ml by UV absorbance at 280 nm
Cross adsorption none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR